Skip to Content
Merck
All Photos(4)

Key Documents

HPA038891

Sigma-Aldrich

Anti-HDC antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Histidine decarboxylase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

PALTSWPSLLGDMLADAINCLGFTWASSPACTELEMNVMDWLAKMLGLPEHFLHHHPSSQGGGVLQSTVSESTLIALLAARKNKILEMKTSEPDADESCLNARLVAYASDQAHSSVEK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDC(3067)

General description

Histidine decarboxylase (HDC) gene codes for acid L-histidine decarboxylase. HDC gene with 14 exons varying in size up to 24 kb is located on human chromosome 15q21–q22.

Immunogen

histidine decarboxylase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Histidine decarboxylase (HDC) is the rate-limiting enzyme required for the synthesis of histamine. Polymorphisms in the HDC gene are linked with abnormal low age at natural menopause (AANM). Female infertility due to ovarian aging is linked to AANM. Obesity, smoking, alcohol usage, and breastfeeding are some of the lifestyle factors linked to AANM.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70111

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleonora Veglia et al.
Pharmacological research, 114, 27-38 (2016-11-05)
Histamine has been reported to decrease the ultrafiltration coefficient, which inversely correlates with glomerular permselectivity, however the mechanism(s) underling this effect have never been investigated. This study aimed to assess whether histamine could exert a direct detrimental effect on podocyte
Feng Zhang et al.
Biochemical and biophysical research communications, 348(4), 1378-1382 (2006-08-22)
Histidine decarboxylase gene (HDC) encodes histidine decarboxylase which is the crucial enzyme for the biosynthesis of histidine. Studies have shown that histamine is likely to be involved in the regulation of reproduction system. To find the possible correlation between HDC
He Dong et al.
PloS one, 11(8), e0160265-e0160265 (2016-08-17)
Tourette syndrome (TS) is a polygenic neuropsychiatric disease. Previous studies have indicated that dysregulation in the histaminergic system may play a crucial role in disease onset. In this study, we investigated the role of the histidine decarboxylase gene (HDC) in
Yoshihiro Inami et al.
Biochemical and biophysical research communications, 535, 19-24 (2020-12-20)
Histamine-producing cells include storage-type cells (e.g., mast cells and basophils), which store histamine intracellularly, and inducible-type cells (e.g., keratinocytes and macrophages), which induce histidine decarboxylase (HDC, a key enzyme for histamine biosynthesis) activity but do not have a storage pool
Alessandro Pini et al.
Pharmacological research, 128, 18-28 (2018-01-09)
Due to the incidence of diabetes and the related morbidity of diabetic nephropathy, identification of new therapeutic strategies represents a priority. In the last few decades new and growing evidence on the possible role of histamine in diabetes has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service