Skip to Content
Merck
All Photos(3)

Documents

HPA029046

Sigma-Aldrich

Anti-PPP2R5D antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2

Synonym(s):

Anti-B56D, Anti-protein phosphatase 2, regulatory subunit B′, δ isoform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PKVAKCTAKPSSSGKDGGGENTEEAQPQPQPQPQPQAQSQPPSSNKRPSNSTPPPTQLSKIKYSGGPQIVKKERRQSSSRFNLSKNRELQKLPAL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP2R5D(5528)

General description

PPP2R5D (protein phosphatase 2 regulatory subunit B′ δ) is a member of the largest PP2A B′ (Protein phosphatase 2A) (B56) subunit family. It codes for B56δ, a 602 amino acid protein, that is a regulatory subunit B of PP2A. In human and mouse, it is more expressed in the progressing brain and less in heart and skeletal muscles.

Immunogen

protein phosphatase 2, regulatory subunit B', delta isoform recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PPP2R5D (protein phosphatase 2 regulatory subunit B′ δ) plays a major role in modulating neuronal and developmental regulation processes such as PI3K (phosphoinositide 3-kinase) /AKT (AKT serine/threonine kinase) and GSK3β (glycogen synthase kinase 3 β) mediated cell growth, chromatin remodeling and gene transcriptional regulation. It also play a vital role in the modulation of Cdc25C (cell division cycle 25C) and Cdk1 (cyclin dependent kinase 1), controlling exit from mitosis. It even controls the dephosphorylation of DARPP-32 (dopamine- and cAMP-regulated phosphoprotein, 32kd; PPP1R1B), thereby modulating the dopaminergic neurotransmission in neurons. Mutations in the PP2A regulatory subunit can cause human overgrowth.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77387

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

De novo missense variants in PPP2R5D are associated with intellectual disability, macrocephaly, hypotonia, and autism
Shang L, et al.
Neurogenetics, 17(1), 43-49 (2016)
Mutations in the PP2A regulatory subunit B family genes PPP2R5B, PPP2R5C and PPP2R5D cause human overgrowth
Loveday C, et al.
Human Molecular Genetics, 24(17), 4775-4779 (2015)
Jade J Dyson et al.
FASEB bioAdvances, 4(4), 273-282 (2022-04-14)
Protein phosphatase 2A (PP2A) is a heterotrimeric phosphatase that controls a wide range of cellular functions. The catalytic activity and intracellular location of PP2A are modulated by its association with regulatory B subunits, including B56 proteins, which are encoded by
Prajakta Varadkar et al.
Cell cycle (Georgetown, Tex.), 16(12), 1210-1219 (2017-06-01)
The Spindle Assembly Checkpoint (SAC) is part of a complex feedback system designed to ensure that cells do not proceed through mitosis unless all chromosomal kinetochores have attached to spindle microtubules. The formation of the kinetochore complex and the implementation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service