Skip to Content
Merck
All Photos(1)

Documents

HPA008840

Sigma-Aldrich

Anti-DUSP12 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dual specificity protein phosphatase 12, Anti-Dual specificity tyrosine phosphatase YVH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

YQAMGYEVDTSSAIYKQYRLQKVTEKYPELQNLPQELFAVDPTTVSQGLKDEVLYKCRKCRRSLFRSSSILDHREGSGPIAFAHKRMTPSSMLTTGRQAQCTSYFIEPVQWMESALLGVMDGQLLCPKCSAKLGSFNWYGEQCSCGRWI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DUSP12(11266)

General description

DUSP12 (dual specificity phosphatase 12) gene is localized to human chromosome 1q21-q22. Its C-terminal contains a non-catalytic domain, which is rich in cysteine and acts as a zinc finger domain. It is an atypical DUSP member, which is recognized as a phosphatase with pro-survival attributes. It is evolutionary conserved. Its DUSP catalytic domain is present at its N-terminal.

Immunogen

Dual specificity protein phosphatase 12 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DUSP12 (dual specificity phosphatase 12) enzyme is thought to be involved in cell survival and the synthesis of ribosomes. It also regulates the progression of cell cycle through its C-terminal zinc-binding domain. It protects cells from oxidative stress, where it forms intramolecular disulfide bonds to prevent irreversible inactivation and remains functionally active. This protein functions as an oncogene, where its over-expression results in elevated cell motility and prevention of apoptosis. It is also lined to metastasis, as its up-regulation leads to elevated levels of c-met proto-oncogene and the collagen and laminin receptor intergrin α1 (itgα1).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70680

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Muda et al.
The Journal of biological chemistry, 274(34), 23991-23995 (1999-08-14)
A human orthologue of the Saccharomyces cerevisiae YVH1 protein-tyrosine phosphatase is able to rescue the slow growth defect caused by the disruption of the S. cerevisiae YVH1 gene. The human YVH1 gene is located on chromosome 1q21-q22, which falls in
Anna Kozarova et al.
Cell cycle (Georgetown, Tex.), 10(10), 1669-1678 (2011-04-28)
The dual-specificity phosphatase hYVH1 (DUSP12) is an evolutionary conserved phosphatase that also contains a unique zinc-binding domain. Recent evidence suggests that this enzyme plays a role in cell survival and ribosome biogenesis. Here, we report that hYVH1 expression also affects
Erica L Cain et al.
PloS one, 6(4), e18677-e18677 (2011-05-11)
Analysis of chromosomal rearrangements within primary tumors has been influential in the identification of novel oncogenes. Identification of the "driver" gene(s) within cancer-derived amplicons is, however, hampered by the fact that most amplicons contain many gene products. Amplification of 1q21-1q23
Christopher A Bonham et al.
The Journal of biological chemistry, 284(34), 22853-22864 (2009-07-02)
YVH1 was one of the first eukaryotic dual specificity phosphatases cloned, and orthologues poses a unique C-terminal zinc-coordinating domain in addition to a cysteine-based phosphatase domain. Our recent results revealed that human YVH1 (hYVH1) protects cells from oxidative stress. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service