Skip to Content
Merck
All Photos(15)

Key Documents

HPA008001

Sigma-Aldrich

Anti-LRPAP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Alpha-2- MRAP, Anti-Alpha-2-macroglobulin receptor-associated protein precursor, Anti-Low density lipoprotein receptor-related protein-associated protein 1, Anti-RAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

RMEKLNQLWEKAQRLHLPPVRLAELHADLKIQERDELAWKKLKLDGLDEDGEKEARLIRNLNVILAKYGLDGKKDARQVTSNSLSGTQEDGLDDPRLEKLWHKAKTSGKFSGEELDKLWREFLHHKEK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRPAP1(4043)

Immunogen

Alpha-2-macroglobulin receptor-associated protein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-LRPAP1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

LRPAP1 (low density lipoprotein receptor-related protein associated protein 1) gene encodes a protein that interacts with Gp330, a low density lipoprotein (LDL) receptor-related protein, and facilitates its proper folding and localization. Gp330 mediates endocytosis of ligands, such as complexes of urokinase and the serpin, plasminogen activator inhibitor-1. LRPAP1 gene is mapped to human chromosome 4p16.3, a region associated with Wolf-Hirschhorn syndrome and disorders of lipoprotein metabolism. It binds to α2-macroglobulin (α2M), a protein that associates with the β-amyloid protein (βA), and functions in the amyloidogenic pathway. Defects in this gene are associated with the risk of developing late-onset Alzheimer disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86748

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Sánchez et al.
American journal of medical genetics, 105(1), 76-78 (2001-06-27)
The LRP-associated protein is involved in the amount of mature LRP expressed on liver and brain. LRP is the main ApoE receptor and also binds alpha2-macroglobulin (alpha2M), a protein that associates with the beta-amyloid protein (betaA). By binding to alpha2M
Lorenz Thurner et al.
Leukemia, 33(1), 148-158 (2018-06-30)
The predominant usage of VH4-34 and V3-21 and reports of stereotyped CDR3s suggest a shared antigenic target of B-cell receptors (BCR) from mantle cell lymphomas (MCL). To identify the target antigens of MCL-BCRs, BCRs from 21 patients and seven MCL
J R Korenberg et al.
Genomics, 22(1), 88-93 (1994-07-01)
Glycoprotein 330 (gp330) is a member of a family of receptors with structural similarities to the low-density lipoprotein receptor. Gp330 is expressed by a number of specialized epithelia, including renal proximal tubules, where it can mediate endocytosis of ligands such
Jan Mulder et al.
Molecular & cellular proteomics : MCP, 8(7), 1612-1622 (2009-04-09)
A need exists for mapping the protein profiles in the human brain both during normal and disease conditions. Here we studied 800 antibodies generated toward human proteins as part of a Human Protein Atlas program and investigated their suitability for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service