Skip to Content
Merck
All Photos(3)

Documents

HPA000804

Sigma-Aldrich

Anti-GRK3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ADRBK2, Anti-BARK2, Anti-β-ARK-2 antibody produced in rabbit, Anti-β-Adrenergic receptor kinase 2 antibody produced in rabbit, Anti-G-protein-coupled receptor kinase 3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

YEAVNADTDKIEARKRAKNKQLGHEEDYALGKDCIMHGYMLKLGNPFLTQWQRRYFYLFPNRLEWRGEGESRQNLLTMEQILSVEETQIKDKKCILFRIKGGKQFVLQCESDPEFVQWKKELNETFKEAQRLLRRAPKFLNKPRS

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADRBK2(157)

Immunogen

G protein-coupled receptor kinase 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

β-adrenergic receptor kinase 2 (β-ARK-2) is an enzyme encoded by the ADRBK2 gene in humans and is mapped to chromosome 22q11. It is a negative regulator of cell growth. ADRBK2 stimulates angiogenesis and is found to be overexpressed in human prostate cancers, especially in metastatic tumors. It plays an important role in prostate cancer progression and metastasis. ADRBK2 is implicated as a candidate molecule for bipolar disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73393

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenliang Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(4), 1521-1526 (2014-01-18)
The biochemical mechanisms that regulate the process of cancer metastasis are still poorly understood. Because kinases, and the signaling pathways they comprise, play key roles in regulation of many cellular processes, we used an unbiased RNAi in vitro screen and
B Mark Woerner et al.
Molecular cancer research : MCR, 10(1), 156-166 (2011-11-17)
G-protein-coupled receptor kinases (GRK) regulate the function of G-protein-coupled receptors (GPCR). Previously, we found that GPCR (CXCR4)-mediated astrocytoma growth was dependent upon abnormally sustained CXCR4 signaling and was correlated with decreased GRK-mediated receptor phosphorylation. As CXCR4 has also been implicated
G Calabrese et al.
Genomics, 23(1), 286-288 (1994-09-01)
Two types of proteins play a major role in determining homologous desensitization of G-coupled receptors: beta-adrenergic receptor kinase (beta ARK), which phosphorylates the agonist-occupied receptor and its functional cofactor, beta-arrestin. Both beta ARK and beta-arrestin are members of multigene families.
Michael J McCarthy et al.
Journal of psychiatric research, 44(4), 201-208 (2009-09-22)
G-protein coupled receptor kinase-3 (GRK3), translated from the gene, ADRBK2 has been implicated as a candidate molecule for bipolar disorder through multiple, converging lines of evidence. In some individuals, the ADRBK2 gene harbors the A-haplotype, a collection of single nucleotide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service