Documents
F9017
SAFC
Flow adapter for jacketed LC columns
column I.D. 1.0 cm
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
column I.D.
1.0 cm
General description
Each flow adapter is supplied with a 20 μm porosity HDPE bed support, TFE/polypropylene O-ring seal, stainless steel adjusting rod, PTFE body and 5 feet (1.5 meters) of FEP tubing.