Key Documents
EP5392070038
Eppendorf(R) Plate Adapter HeatSealer S200
for 96 and 384 well plates
Select a Size
1 EA
€375.00
€375.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€375.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.15
€375.00
Please contact Customer Service for Availability
feature
for 96 and 384 well plates
manufacturer/tradename
Eppendorf® 5392070038 (Global)
Related Categories
Features and Benefits
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)