Skip to Content
Merck
All Photos(1)

Documents

EHU068241

Sigma-Aldrich

MISSION® esiRNA

targeting human P2RX4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATACACGGGACGTTGAGCACAACGTATCTCCTGGCTACAATTTCAGGTTTGCCAAGTACTACAGAGACCTGGCTGGCAACGAGCAGCGCACGCTCATCAAGGCCTATGGCATCCGCTTCGACATCATTGTGTTTGGGAAGGCAGGGAAATTTGACATCATCCCCACTATGATCAACATCGGCTCTGGCCTGGCACTGCTAGGCATGGCGACCGTGCTGTGTGACATCATAGTCCTCTACTGCATGAAGAAAAGACTCTACTATCGGGAGAAGAAATATAAATATGTGGAAGATTACGAGCAGGGTCTTGCTAGTGAGCTGGACCAGTGAGGCCTACCCCACACCTGGGCTCTCCACAGCCCCATCAAAGAACAGAGAGGAGGAGGAGGGAGAAATGGCCACCACATCACCCCAGAGAAATTTCTGGAATCTGATTGAGTCTCCACTCCACAAGCACTCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Feng Huo et al.
Journal of cellular biochemistry, 120(4), 6322-6329 (2018-10-27)
Purinergic receptor P2X 4 (P2X4R), a member of purinergic channels family and a subtype of ionotropic adenosine triphosphate receptors, plays a critical role in tumorigenesis. Evidence suggested that P2X4R is expressed in rat C6 glioma model, however, its role and
Dmitry Aminin et al.
Scientific reports, 6, 39683-39683 (2016-12-23)
Since ancient times, edible sea cucumbers have been considered a jewel of the seabed and used in Asian folk medicine for stimulation of resistance against different diseases. However, the power of this sea food has not been established on a
Larisa Gofman et al.
Alcohol and alcoholism (Oxford, Oxfordshire), 51(6), 647-654 (2016-10-30)
Previously we have demonstrated altered microglia P2X4R expression in response to alcohol and pharmacological blockade with a selective P2X4R antagonist can reverse the action, suggesting that P2X4R play a role in mediating alcohol-induced effects on microglia. In the present study
Xiao-Hong Jin et al.
Journal of neuroscience research, 92(12), 1690-1702 (2014-07-06)
Previous studies have suggested that the microglial P2X7 purinoceptor is involved in the release of tumor necrosis factor-α (TNFα) following activation of toll-like receptor-4 (TLR4), which is associated with nociceptive behavior. In addition, this progress is evoked by the activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service