Skip to Content
Merck
All Photos(1)

Key Documents

MAT-LyLu

94101454, rat prostate, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102

product name

MAT-LyLu, 94101454

biological source

rat prostate

growth mode

Adherent

karyotype

Modal no. 64

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat prostate adenocarcinoma

Cell Line Description

A rat prostate tumour cell line derived from successive in vivo passages of the AT-1 tumour cell line (Sigma Catalogue number. 94101449). The cells are anaplastic, androgenin dependant and metastatic to the lung.

Culture Medium

RPMI 1640 + 2mM Glutamine + 250nM Dexamethasone (DXMT) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:20 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service