Skip to Content
Merck
All Photos(1)

Key Documents

Fc3Tg

90073002, feline embryonic tongue. Fibroblast-like morphology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Fc3Tg, 90073002

biological source

feline mouth

growth mode

Adherent

karyotype

2n = 38

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Cat tongue

Cell Line Description

Established from normal embryonal tongue tissue of female cat (Felis catus). Cells support the replication of a wide range of feline viruses. Can undergo 50 population doublings.

Application

Virus replication studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2 37°C. Maintain at pH 7.1-7.3.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service