Skip to Content
Merck
All Photos(1)

Documents

RD

85111502, human embryo, Spindle shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RD, 85111502

biological source

human embryo

growth mode

Adherent

karyotype

Modal no. 56

morphology

Spindle shaped

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian embryo rhabdomyosarcoma

Cell Line Description

Derived from a malignant embryonal rhabdomyosarcoma from a 7 year old Caucasian female. Also designated TE 32 and 130T in the literature

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 13
D16S539: 10,11
D5S818: 11
D7S820: 8,12
THO1: 9.3
TPOX: 9
vWA: 18

Culture Medium

DMEM + 2mM Glutamine + 2% Non Essential Amino Acids (NEAA) + 2% Vitamins + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service