Skip to Content
Merck
All Photos(1)

Key Documents

C3366

Sigma-Aldrich

Anti-CXCR5 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CXC Chemokine Receptor 5

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€557.00

€557.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μL
€557.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

€557.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5 μg/mL using human lymph node, B-cell lymphoma

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXCR5(643)
mouse ... Cxcr5(12145)
rat ... Cxcr5(29363)

Immunogen

synthetic peptide corresponding to the C-terminus of human CXC chemokine receptor 5 (CXCR5), conjugated to KLH. The immunizing peptide has 94% homology with the rat gene and 100% homology with the mouse gene.

Biochem/physiol Actions

CXCR5 is a multi-pass membrane protein expressed in mature B cells. It binds to B-lymphocyte chemoattractant and regulates the migration of B cells. The expression of CXCR5 is important for CD4 T cell-mediated humoral immune responses.

Physical form

Solution in PBS, 0.1% Sodium Azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Through the looking glass: the diverse in vivo activities of chemokines.
S L Kunkel
The Journal of clinical investigation, 104(10), 1333-1334 (1999-11-24)
Nina Chevalier et al.
Journal of immunology (Baltimore, Md. : 1950), 186(10), 5556-5568 (2011-04-08)
High expression of CXCR5 is one of the defining hallmarks of T follicular helper cells (T(FH)), a CD4 Th cell subset that promotes germinal center reactions and the selection and affinity maturation of B cells. CXCR5 is also expressed on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service