Skip to Content
Merck
All Photos(2)

Documents

AV38028

Sigma-Aldrich

Anti-ELF4 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-E74-like factor 4 (ets domain transcription factor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

71 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELF4(2000)

Related Categories

General description

ETS transcription factors share a conserved DNA-binding "ETS" domain and include several oncoproteins that induce tumorigenesis when overexpressed. The ETS family member E74-like factor 4 (ELF4) controls the ERK-mediated proliferation and homing of CD8+ T cells via Krüppel-like factors KLF4 and KLF2. ELF4 increase quiescence in bone marrow endothelial cells by the deregulation of cyclin-dependent kinase-4 expression and enhances regeneration of sinusoidal blood vessels.

Specificity

Anti-ELF4 (AB1) polyclonal antibody reacts with bovine, human, mouse, rat, and canine E74-like factor 4 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human ELF4

Application

Anti-ELF4 (AB1) polyclonal antibody is used to tag E74-like factor 4 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of E74-like factor 4 in the signaling and regulation of quiescence and proliferation of cells such at T-cells and endothelial cells.

Biochem/physiol Actions

ELF4 contains 1 ETS DNA-binding domain and belongs to the ETS family. It is transcriptional activator that binds to DNA sequences containing the consensus 5′-WGGA-3′. It transactivates promoters of the hematopoietic growth factor genes CSF2, IL3, IL8, and of the bovine lysozyme gene. It acts synergistically with RUNX1 to transactivate the IL3 promoter and also transactivates the PRF1 promoter in natural killer (NK) cells. ELF4 plays a role in the development and function of NK and NK T-cells and in innate immunity.

Sequence

Synthetic peptide located within the following region: MAITLQPSDLIFEFASNGMDDDIHQLEDPSVFPAVIVEQVPYPDLLHLYS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fuping You et al.
Nature immunology, 14(12), 1237-1246 (2013-11-05)
Induction of type I interferon is a central event of innate immunity, essential for host defense. Here we report that the transcription factor ELF4 is induced by type I interferon and upregulates interferon expression in a feed-forward loop. ELF4 deficiency
Lili Cao et al.
Science signaling, 12(573) (2019-03-21)
Precise control of interferons (IFNs) is crucial to maintain immune homeostasis. Here, we demonstrated that homeodomain-interacting protein kinase 2 (HIPK2) was required for the production of type I IFNs in response to RNA virus infection. HIPK2 deficiency markedly impaired IFN

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service