Skip to Content
Merck
All Photos(1)

Documents

A8451

Sigma-Aldrich

(−)-cis,trans-Abscisic acid

Synonym(s):

(R)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl)-3-methyl-(2Z,4E)-pentadienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

Quality Level

application(s)

agriculture

storage temp.

−20°C

SMILES string

CC(\C=C\[C@]1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O

InChI

1S/C15H20O4/c1-10(7-13(17)18)5-6-15(19)11(2)8-12(16)9-14(15,3)4/h5-8,19H,9H2,1-4H3,(H,17,18)/b6-5+,10-7-/t15-/m0/s1

InChI key

JLIDBLDQVAYHNE-QHFMCZIYSA-N

Application

Abscisic acid (ABA) plays an important role in plant growth and development, regulation of seed maturation, germination, and adaptation to environmental stresses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Gampala et al.
The Journal of biological chemistry, 276(13), 9855-9860 (2001-01-15)
cis,trans-Abscisic acid (ABA) plays an important role in plant growth and development, regulation of seed maturation, germination, and adaptation to environmental stresses. Knowledge of ABA mechanisms of action and the interactions of components required for ABA signal transduction is far
Veronika Hradecká et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 847(2), 162-173 (2006-10-27)
Polyclonal antibodies with high specificity for C1-immobilised (+)-cis,trans-abscisic acid (ABA) were raised, characterised by enzyme-linked immunosorbent assay (ELISA) and used for preparation of an immunoaffinity chromatography (IAC) gel. The detection limit of the ELISA was approximately 4.6x10(-10)mol/L. Sensitive electrospray liquid
Carlo I G Tuberoso et al.
Journal of agricultural and food chemistry, 58(1), 384-389 (2009-11-19)
Strawberry tree honey, due to its characteristic bitter taste, is one of the most typical Mediterranean honeys, with Sardinia being one of the largest producers. According to specific chemical studies, homogentisic acid was identified as a possible marker of this
Helene Persak et al.
PloS one, 8(2), e57547-e57547 (2013-02-26)
Abiotic stress poses a huge, ever-increasing problem to plants and agriculture. The dissection of signalling pathways mediating stress tolerance is a prerequisite to develop more resistant plant species. Mitogen-activated protein kinase (MAPK) cascades are universal signalling modules. In Arabidopsis, the
Belmiro Vilela et al.
PloS one, 8(2), e58105-e58105 (2013-03-08)
The Arabidopsis kinase OPEN STOMATA 1 (OST1) plays a key role in regulating drought stress signalling, particularly stomatal closure. We have identified and investigated the functions of the OST1 ortholog in Z. mays (ZmOST1). Ectopic expression of ZmOST1 in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service