A5846
Anemone Toxin II recombinant from Escherichia coli
>95%, lyophilized powder
Synonym(s):
ATX II
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
Assay
>95%
form
lyophilized powder
mol wt
4932
storage temp.
−20°C
Biochem/physiol Actions
Originally isolated from Anemonia sulcata
Voltage-gated sodium channel activator
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service