Skip to Content
Merck
All Photos(15)

Key Documents

ZRB1267

Sigma-Aldrich

Anti-CGRP-1 Antibody, clone 2E18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-CALC, Anti-CCALCI, Anti-CGRP-I

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€191.00
4 X 25 μL
€393.00

€191.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€191.00
4 X 25 μL
€393.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€191.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E18, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~13.90 kDa
observed mol wt ~14 kDa

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2E18 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Calcitonin gene-related peptide 1. It targets an epitope within 22 amino acids from the active form of CGRP1.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from human active Calcitonin gene-related peptide 1 (C-terminal region of the uncleaved peptide)

Application

Quality Control Testing

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Calcitonin Gene-Related Peptide 1 (CGRP-1) in MCF-7 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CGRP-1 in PC12 and K562 cells and in Human and mouse liver lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CGRP-1 in human cerebral cortex and human DRG tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound CGRP-1 peptide with a KD of 1.9 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected CGRP-1 in MCF7 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CGRP-1, clone 2E18 ZooMAb®, Cat. No. ZRB1267, is a recombinant Rabbit monoclonal antibody that specifically targets CGRP-1 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Calcitonin gene-related peptide 1 (UniProt: P06881; also known as Alpha-type CGRP, Calcitonin gene-related peptide I, CGRP-I) is encoded by the CALCA (also known as CALC1) gene (Gene ID: 746) in human. CGRP, a 37 amino acid neuropeptide, is a highly potent and long-acting vasodilator with protective effects in the cardiovascular system. Two forms of CGRP have been described -CGRP-I and CGRP-II (also known as aCGRP and bCGRP) that are synthesized from two distinct genes. CGRP I is the primary neuronal form that is found in the CNS and in the peripheral nervous system and displays higher immunoreactivity than CGRP II. CGRP I is synthesized with a signal peptide (aa 1-25) and two peptides (aa 26-80 and 125-128) that are cleaved off to generate biologically active form (aa 83-119). Following synthesis, it is stored in large, dense-core vesicles within the sensory nerve terminal and is released following neuronal depolarization via calcium-dependent exocytosis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Russell, FA., et al. (2014). Physiol. Rev. 94(4): 1099-1142).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service