Key Documents
S8400
Slides, microscope
frosted one end, size 25 mm × 75 mm
About This Item
UNSPSC Code:
41122600
NACRES:
MA.01
material
glass
packaging
pkg of 1 Case - 20 boxes of 72 slides (1440)
pkg of 1 Pack - box of 72 slides
size
25 mm × 75 mm
binding type
non-treated surface
General description
Sealed in a moisture absorbing desiccant pack which can function as a drying stand and a storage box. Slides are 1 mm thick.
Our microscope slides are made of high-quality white glass with low iron content, no green tint with fine texture and transparency; these slides are also corrosion-resistant. The edges are ground smooth to help eliminate sharp cutting surfaces and safe handling. They are packed in a special thermoform plastic box to minimize exposure to moisture and dust (each slide is precleaned).
Conform to Federal specification NNN-S-450.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Geovanny Nic-Can et al.
Plant methods, 9(1), 47-47 (2013-12-18)
Epigenetic mechanisms can be highly dynamic, but the cross-talk among them and with the genome is still poorly understood. Many of these mechanisms work at different places in the cell and at different times of organism development. Covalent histone modifications
Ons M'Saad et al.
Nature communications, 11(1), 3850-3850 (2020-08-02)
Resolving the distribution of specific proteins at the nanoscale in the ultrastructural context of the cell is a major challenge in fluorescence microscopy. We report the discovery of a new principle for an optical contrast equivalent to electron microscopy (EM)
Articles
Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.
Related Content
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.