L8795
LB Agar Tetracycline-10, Plates
pre-poured agar plates with 10 μg/ml tetracycline
About This Item
Recommended Products
grade
for molecular biology
sterility
sterile; autoclaved
form
plates (10cm, 20-25ml each)
availability
available only in USA
storage temp.
2-8°C
suitability
selective for Escherichia coli
selective for coliforms
General description
Application
Features and Benefits
- Convenient, ready-to-use 10cm format
- Standard formulation plus antibiotic
Components
10g/L Tryptone
10g/L NaCl
5g/L Yeast Extract
10μg/ml tetracycline
Other Notes
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Why do the LB-agar plates with antibiotics have such a short expiration date?
The antibiotics in the LB-agar plates are not stable. The antibiotic degrades over time, making it ineffective at selecting transformed bacteria grown on the plate. Because of this, we recommend the plates are used as soon as possible once they are received. For labs with low use, the plates may expire quickly, so they may benefit from pouring their own plates using the LB - Miller powder with agar formulation (L3147).
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service