Skip to Content
Merck
All Photos(1)

Key Documents

79398

Sigma-Aldrich

1-(3-sn-Phosphatidyl)-rac-glycerol ammonium salt solution

egg yolk(lecithin), ≥99.0% (TLC)

Synonym(s):

L-α-Phosphatidyl-DL-glycerol ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51191904
NACRES:
NA.25

biological source

egg yolk (lecithin)

Assay

≥99.0% (TLC)

form

liquid

composition

16:0, ~36% (fatty acid)
18:0, ~10% (fatty acid)
18:1, ~33% (fatty acid)
18:2, ~14% (fatty acid)

concentration

1% in chloroform: methanol (49:1)

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

wet ice

storage temp.

−20°C

SMILES string

N.OCC(O)COP(O)(=O)OC[C@H](O)CO

Preparation Note

by reaction of egg-yolk PC with cabbage phospholipase D in the presence of glycerol

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service