Skip to Content
Merck
All Photos(1)

Documents

205303

Sigma-Aldrich

Lithium perchlorate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiClO4 · 3H2O
CAS Number:
Molecular Weight:
160.44
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

form

crystals and lumps

reaction suitability

reagent type: oxidant

concentration

21-22% (chlorine basis (silver nitrate titration))

density

1.841 g/mL at 25 °C (lit.)

SMILES string

[Li+].[H]O[H].[H]O[H].[H]O[H].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Li.3H2O/c2-1(3,4)5;;;;/h(H,2,3,4,5);;3*1H2/q;+1;;;/p-1

InChI key

JAWYRNYHJJDXHX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Crystal structure of lithium perchlorate trihydrate (LiClO4.3H2O) has been investigated by X-ray and neutron diffraction studies. It exhibits pyroelectric properties.

Application

Lithium perchlorate trihydrate may be used to modify the membrane morphology of poly[(vinylidene fluoride)-co-trifluoroethylene] membranes, by doping it with varying concentrations of LiClO4.3H2O. It may be used to compose the polymer nanocomposite electrolyte (PNCE) films.
When used as a co-catalyst, accelerates the Baylis-Hillman reaction, the coupling of α,β-unsaturated carbonyls with aldehydes.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of the microsctructure and lithium-ion content in poly [(vinylidene fluoride)-co-trifluoroethylene]/lithium perchlorate trihydrate composite membranes for battery applications.
Costa CM, et al.
Solid State Ionics, 217, 19-26 (2012)
Neutron diffraction refinement of pyroelectric lithium perchlorate trihydrate.
Lundgren JO, et al.
Acta Crystallographica Section B, Structural Crystallography and Crystal Chemistry, 38(1), 15-20 (1982)
The structure of lithium perchlorate trihydrate Li(H2O)3ClO4-an X-ray and neutron diffraction study.
Sequeira A, et al.
Acta Crystallographica Section B, Structural Crystallography and Crystal Chemistry, 31(6), 1735-1739 (1975)
Jian-Yu Lu et al.
Polymers, 13(3) (2021-01-28)
Hydroquinone (HQ) is one of the major deleterious metabolites of benzene in the human body, which has been implicated to cause various human diseases. In order to fabricate a feasible sensor for the accurate detection of HQ, we attempted to
Ionic conduction and relaxation processes in melt compounded poly(ethylene oxide)-lithium perchlorate trihydrate-montmorillonite nanocomposite electrolyte.
Choudhary S and Sengwa RJ.
Indian Journal of Engineering and Materials Sciences, 18(2), 147-156 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service