B49203
Bis(N-methylacridinium) nitrate
97%
Synonym(s):
N,N′-Dimethyl-9,9′-biacridinium dinitrate, Lucigenin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
mp
245-250 °C (dec.) (lit.)
λmax
430 nm
452 nm (2nd)
SMILES string
O[N+]([O-])=O.O[N+]([O-])=O.C[n+]1c2ccccc2c(-c3c4ccccc4[n+](C)c5ccccc35)c6ccccc16
InChI
1S/C28H22N2.2NO3/c1-29-23-15-7-3-11-19(23)27(20-12-4-8-16-24(20)29)28-21-13-5-9-17-25(21)30(2)26-18-10-6-14-22(26)28;2*2-1(3)4/h3-18H,1-2H3;;/q+2;2*-1
InChI key
KNJDBYZZKAZQNG-UHFFFAOYSA-N
Other Notes
This product has been replaced by M8010-Sigma-Aldrich | N,N′-Dimethyl-9,9′-biacridinium dinitrate used as chemiluminescent reagent
replaced by
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Determination of conjugated glucuronic acid by combining enzymatic hydrolysis with lucigenin chemiluminescence.
Analytical chemistry, 57(1), 46-51 (1985-01-01)
Analytical and bioanalytical chemistry, 401(1), 381-386 (2011-05-17)
The bright chemiluminescence has been observed in the system: Co(2+)/hydrogen peroxide/lucigenin. The chemiluminescence intensity was directly proportional to either cobalt, hydrogen peroxide, or lucigenin concentrations. A procedure of determination of superoxide dismutase (SOD) activity by the chemiluminescence method in the
Analytical biochemistry, 395(2), 134-143 (2009-08-15)
NAD(P)H oxidoreductases of the outer mitochondrial membrane (OMM) are able to activate various xenobiotics and stimulate the production of reactive oxygen species and the opening of the mitochondrial permeability transition pore. However, the role of these systems in the cell
Journal of clinical periodontology, 39(7), 626-634 (2012-05-23)
To determine the effect of cigarette smoke extract, nicotine and cotinine on lucigenin-detectable neutrophil superoxide production. Neutrophils from periodontally healthy individuals were treated with aqueous smoke extract, nicotine and cotinine, prior to stimulation or at the same time as stimulation
Chemistry (Weinheim an der Bergstrasse, Germany), 18(16), 4823-4826 (2012-03-16)
Multifunctional hybrid: A dendritic platinum nanoparticle/lucigenin/reduced graphene oxide (RGO) hybrid with chemiluminescence (CL) activity was prepared for the first time by using lucigenin as a linker through simultaneous reduction of H(2) PtCl(4) and a lucigenin-functionalized graphene oxide composite by sodium
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service