Skip to Content
Merck
All Photos(1)

Documents

86181

Sigma-Aldrich

10-(3-Sulfopropyl)acridinium betaine

purum, for fluorescence, ≥98.0% (HPLC)

Synonym(s):

3-(10-Acridinio)propanesulfonate, N-(3-Sulfopropyl)acridinium inner salt, SPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15NO3S
CAS Number:
Molecular Weight:
301.36
Beilstein:
4555235
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence
purum

Assay

≥98.0% (HPLC)

mp

280-290 °C (dec.)

fluorescence

λex 415 nm; λem 488 nm in H2O

SMILES string

[O-]S(=O)(=O)CCC[n+]1c2ccccc2cc3ccccc13

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Standard for the determination of fluorescence lifetimes; Determination of various species by fluorescence quenching

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Trettnak et al.
Fresenius Journal of Analytical Chemistry, 326, 547-547 (1987)
O.S. Wolfbeis et al.
Fresenius Journal of Analytical Chemistry, 314, 577-577 (1983)
Phase fluorometric method for determination of standard lifetimes.
R B Thompson et al.
Analytical chemistry, 60(7), 670-674 (1988-04-01)
R Krapf et al.
Analytical biochemistry, 169(1), 142-150 (1988-02-15)
The application of the quinoline derivative 6-methoxy-N-(3-sulfopropyl)quinolinium (SPQ) to the measurement of membrane transport of chloride in biological systems was reported recently (N.P. Illsley and A.S. Verkman (1987) Biochemistry 26, 1215-1219). To understand the structure-activity relationships of compounds with chloride-sensitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service