Skip to Content
Merck
All Photos(1)

Documents

84720

Sigma-Aldrich

Sulfuric acid

puriss. p.a., 95-97% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

vapor density

<0.3 (25 °C, vs air)

Quality Level

vapor pressure

1 mmHg ( 146 °C)

description

Nominally 95-98% H2SO4

grade

puriss. p.a.

Assay

95-97% (T)

form

liquid

availability

not available in (Sales restrictions may apply)

impurities

≤0.0002% KMnO4-red. substances (as SO2)

ign. residue

≤0.0005%

color

clear

bp

~290 °C (lit.)

density

1.83 g/mL at 20 °C
1.840 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.1 mg/kg
nitrate (NO3-): ≤0.2 mg/kg
phosphate (PO43-): ≤0.5 mg/kg

cation traces

Ag: ≤0.02 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.01 mg/kg
Ba: ≤0.05 mg/kg
Be: ≤0.01 mg/kg
Bi: ≤0.05 mg/kg
Ca: ≤0.2 mg/kg
Cd: ≤0.02 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.05 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.1 mg/kg
Ge: ≤0.05 mg/kg
Hg: ≤0.005 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.05 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.02 mg/kg
NH4+: ≤2 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.02 mg/kg
Sr: ≤0.02 mg/kg
Ti: ≤0.1 mg/kg
Tl: ≤0.05 mg/kg
V: ≤0.01 mg/kg
Zn: ≤0.05 mg/kg
Zr: ≤0.1 mg/kg

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

In PE-bottles; also available Glass-bottles (Material no. ending with *-GL)

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
Sales restrictions may apply

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of mercury in natural waters and effluents by flameless atomic absorption spectrophotometry.
S H Omang
Analytica chimica acta, 53(2), 415-419 (1971-02-01)
Jing Li et al.
Journal of chromatography. A, 1404, 39-43 (2015-06-09)
The paper reports on a novel method for rapid determination of carbonyl in lignins by headspace gas chromatography (HS-GC). The method involves the quantitative carbonyl reduction for aldehydes in 2min at room temperature or for acetones in 30min at 80°C
Investigation of the kinetics and mechanism of the anodic oxidation of aniline in aqueous sulfuric acid solution at a platinum electrode.
Mohilner DM, et al.
Journal of the American Chemical Society, 84(19), 3618-3622 (1962)
Self-Ordering of Cell Arrangement of Anodic Porous Alumina Formed in Sulfuric Acid Solution.
Masuda H, et al.
Journal of the Electrochemical Society, 144(5), L127-L130 (1997)
João Almeida et al.
Nature, 502(7471), 359-363 (2013-10-08)
Nucleation of aerosol particles from trace atmospheric vapours is thought to provide up to half of global cloud condensation nuclei. Aerosols can cause a net cooling of climate by scattering sunlight and by leading to smaller but more numerous cloud

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service