Skip to Content
Merck
All Photos(3)

Documents

67447

Millipore

STEC Millichrome plus Agar Base

for bacteria: Escherichia coli (Shiga toxin STEC), suitable for microbiology

Synonym(s):

Agar Base for STEC, Chromogenic Agar Base for STEC Detection, STEC Detection and Identification Agar Base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161501
NACRES:
NA.74

Agency

EP
JP
USP

Quality Level

Sterilization Compatibility

dry heat compatible

form

dry powder

shelf life

limited shelf life, expiry date on the label

composition

Agar, 15.0 g/L
Chromogenic Mix, 2.6 g/L
Peptone & Yeast extract, 8.0 g/L
Salts, 5.2 g/L

manufacturer/tradename

MilliChrome

technique(s)

microbiological culture: suitable

final pH

6.0±0.2 (25 °C)

solubility

deionized water: 30.8 g/L

application(s)

food and beverages
microbiology

shipped in

ambient

storage temp.

15-25°C

suitability

bacteria: Escherichia coli (Shiga toxin STEC)

General description

STEC Millichrome plus Agar Base is a selective chromogenic culture medium used for the identification of Shiga-like toxin-producing E. coli (STEC). The media contains chromogenic substrates that produce visible color changes upon enzymatic activity of E. coli. Peptone and yeast extract are nitrogenous nutrients for growth and other essential growth factors. Sodium chloride provides osmotic balance. The chromogenic mix contains chromogenic substrates for color differentiation based on the ability to cleave the substrate by characteristic enzymes. STEC Millichrome plus supplement contains selective agents that allow STEC to grow while other organisms are inhibited. Agar is a solidifying agent.

Application

STEC Millichrome plus Agar Base is a highly selective chromogenic culture medium intended for use in the qualitative direct detection, differentiation, and presumptive identification of Shiga-like-toxin producing E. coli (STEC) in various samples, including food products for human consumption, animal feed, and environmental samples.

Preparation Note

Step 1 (Preparation of the base STEC Millichrome plus Agar Base)

  • Disperse slowly 30.8 g of powder base in 1 L of purified water.
  • Stir until agar is well thickened.
  • Heat and bring to boil (100 °C) while swirling or stirring regularly. DO NOT HEAT TO MORE THAN 100 °C. DO NOT AUTOCLAVE AT 121 °C.
Warning: If using an autoclave, do so without pressure. Advice 1: For the 100 °C heating step, mixture may also be brought to a boil in a microwave oven: after initial boiling, remove from oven, stir gently, then return to oven for short repeated bursts of heating until complete fusion of the agar grains has taken place (large bubbles replacing foam).
  • Cool in a water bath to 45-50 °C. Swirl or stir gently to homogenize.
Step 2 (Preparation of the STEC Millichrome plus Agar Base and mix with base)
  • Aseptically rehydrate 1 vial with 10 mL of sterile water.
  • Swirl well until complete dissolution.
  • Add 1 vial rehydrated solution to 1L STEC Millichrome plus Agar Base cooled at 45-50 °C.
  • Swirl gently to homogenize.
Step 3 Pour plates
  • Pour into sterile Petri dishes
  • Let it solidify and dry.

Storage and Stability

  • Store prepared media below 8°C, protected from direct light (max. 1 month).
  • Store dehydrated powder, in a dry place, in tightly sealed containers at 2-25°C.
  • The rehydrated supplement is stable for up to 2 months when stored at 2-8°C

Analysis Note

The presence of mauve colonies indicates the presence of STEC. Other organisms may show colorless or blue colonies or be inhibited. STEC O157 is nonfluorescent under UV light, while other STEC serotypes are variable in fluorescence under the UV lamp at 365nm.

Legal Information

MilliChrome is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The product is notintended for diagnosing infections or guiding treatment.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service