Skip to Content
Merck
All Photos(1)

Documents

612081

Supelco

Gadolinium(III) chloride solution

NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), DSS 0.1 %, water 1 %, NMR tube size 3 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GdCl3
CAS Number:
Molecular Weight:
263.61
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

NMR reference standard
analytical standard

Quality Level

composition

DSS, 0.1%
water, 1%

concentration

0.1 mg/mL in D2O (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

format

single component solution

SMILES string

Cl[Gd](Cl)Cl

InChI

1S/3ClH.Gd/h3*1H;/q;;;+3/p-3

InChI key

MEANOSLIBWSCIT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Gradient shimming

Quantity

3 mm O.D. tube contains 0.235 mL.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quirin Zangl et al.
Anesthesiology, 121(6), 1217-1225 (2014-09-17)
Liver damage by ischemia and reperfusion injury is a risk factor for morbidity and mortality after liver surgery. Postoperative oxygen treatment is routinely applied in the postanesthesia and intensive care unit after liver surgery. The risks of aggravating the injury
Yan Yang et al.
European journal of immunology, 44(8), 2468-2477 (2014-04-30)
Basophils have been shown to contribute to anaphylaxis through either an IgE-FcεRI-dependent pathway or an IgG-FcγR pathway. However, it remains largely unclear whether basophils can be activated to promote anaphylaxis via a non-FcR pathway as well. The glycolipid receptor ASGM1
Christopher G J McKenzie et al.
Blood, 123(22), 3496-3503 (2014-03-19)
Transfusion-related acute lung injury (TRALI) is the leading cause of transfusion-related mortality and can occur with any type of transfusion. TRALI is thought to be primarily mediated by donor antibodies activating recipient neutrophils resulting in pulmonary endothelial damage. Nonetheless, details
Hiroaki Kito et al.
Biochemical and biophysical research communications, 459(3), 457-462 (2015-03-10)
Store-operated Ca(2+) entry (SOCE) via Orai1 and STIM1 complex is supposed to have obligatory roles in the regulation of cellular functions of vascular endothelial cells, while little is known about the contribution of Orai2. Quantitative PCR and Western blot analyses
Alfredo Mijares et al.
Biochemical and biophysical research communications, 452(4), 1054-1059 (2014-09-23)
Duchenne muscular dystrophy (DMD) is a lethal X-inherited disease caused by dystrophin deficiency. Besides the relatively well characterized skeletal muscle degenerative processes, DMD is also associated with a dilated cardiomyopathy that leads to progressive heart failure at the end of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service