Skip to Content
Merck
All Photos(1)

Key Documents

428760

Supelco

Amberlite HPR9200 Ion Exchange Resin

chloride form, 25-30 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

product name

AmberLite HPR9200 Cl, chloride form, chloride form

Quality Level

description

formerly Dowex Marathon MSA Cl

form

beads

moisture

60-72%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (macroporous)

matrix active group

quaternary amine

particle size

23-27 mesh
560-660 μm

capacity

1.0 meq/mL by wetted bed volume

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Dowex Marathon MSA is a strong anion exchanger resin, with a type I quaternary ammonium functional group, macroporous structure and monodisperse bead size. This resin is opted due to its capacity and catalytic activity.

Application

Dowex Marathon MSA was used to determine the catalytic activity under mild temperature conditions.
Strongly basic (type I) anion exchange resin for removal of organic impurities with minimal organic fouling. Also useful for deionization, silica removal, and condensate polishing. this resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Other Notes

The Dowex Marathon MSA is a superior resin to the Dowex MSA-1. The Dowex Marathon series provides narrower particle distribution, better efficiency, and improved kinetics when compared to the gaussian Dowex products.
This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Green upgrading of succinate using dimethyl carbonate for a better integration with fermentative production.
Lopez-Garzon, Camilo S., Luuk AM van der Wielen, and Adrie JJ Straathof.
Chemical Engineering Journal, 235, 52-60 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service