Skip to Content
Merck
All Photos(1)

Documents

07-6140

Sigma-Aldrich

Dithizone

JIS special grade, ≥85.0%

Synonym(s):

Diphenylthiocarbazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNHCSN=NC6H5
CAS Number:
Molecular Weight:
256.33
Beilstein:
748838
EC Number:
MDL number:
UNSPSC Code:
12352127
PubChem Substance ID:

grade

JIS special grade

Assay

≥85.0%

form

powder

availability

available only in Japan

mp

168 °C (dec.) (lit.)

SMILES string

S=C(NNc1ccccc1)\N=N\c2ccccc2

InChI

1S/C13H12N4S/c18-13(16-14-11-7-3-1-4-8-11)17-15-12-9-5-2-6-10-12/h1-10,14H,(H,16,18)/b17-15+

InChI key

UOFGSWVZMUXXIY-BMRADRMJSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaodong Wen et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 782-787 (2012-08-21)
In this work, a new coupling of spectrophotometric determination with preconcentration method named as ultrasound-assisted emulsification dispersive liquid-liquid microextraction (UAE-DLLME) for trace silver was firstly established. Disperser solvent in traditional DLLME was substituted by ultrasound-assisted emulsification, which could afford more
Jianjun Deng et al.
Journal of hazardous materials, 196, 187-193 (2011-09-29)
An electrospun nanofiber was utilized as a sorbent in packed fiber solid phase extraction (PFSPE) for selective separation and preconcentration of lead (II). The nanofiber had a polystyrene (PS) backbone, which was functionalized with dithizone (DZ) by co-electrospinning of a
Anna Pisania et al.
Laboratory investigation; a journal of technical methods and pathology, 90(11), 1661-1675 (2010-08-11)
Despite improvements in outcomes for human islet transplantation, characterization of islet preparations remains poorly defined. This study used both light microscopy (LM) and electron microscopy (EM) to characterize 33 islet preparations used for clinical transplants. EM allowed an accurate identification
Yukun Wang et al.
Analytica chimica acta, 716, 112-118 (2012-01-31)
Graphene, a novel class of carbon nanostructures, has great promise for use as sorbent materials because of its ultrahigh specific surface area. A new method using a column packed with graphene as sorbent was developed for the preconcentration of trace
A Słomka et al.
Journal of plant physiology, 168(11), 1191-1199 (2011-04-16)
Violets of the sections Melanium were examined for their colonization by arbuscular mycorrhizal fungi (AMF). Heartsease (Viola tricolor) from several heavy metal soils was AMF-positive at many sites but not at extreme biomes. The zinc violets Viola lutea ssp. westfalica

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service