Skip to Content
Merck
All Photos(1)

Documents

05-6490

Sigma-Aldrich

m-Cresol

SAJ first grade, ≥97.0%

Synonym(s):

1-Hydroxy-3-methylbenzene, 3-Hydroxytoluene, 3-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
506719
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.72 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥97.0%

autoignition temp.

1036 °F

expl. lim.

1.06-1.35 %, 150 °F

availability

available only in Japan

refractive index

n20/D 1.541 (lit.)

bp

203 °C (lit.)

mp

8-10 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(O)c1

InChI

1S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8H,1H3

InChI key

RLSSMJSEOOYNOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Saravanan et al.
Bioresource technology, 99(18), 8553-8558 (2008-05-13)
An internal loop airlift reactor (ILALR) is developed and studied for biodegradation of phenol/m-cresol as single and dual substrate systems under batch and fed batch operation using an indigenous mixed microbial strain, predominantly Pseudomonas sp. The results showed that the
Chiu-Po Chan et al.
Toxicology, 208(1), 95-104 (2005-01-25)
Cresol is a well-known environmental pollutant, toluene metabolite, uremic toxicant and accidental poisoning product. Formocresol, a preparation of formalin and cresol, is also used as a root canal medicament and for pulpotomy of primary teeth. However, little is known about
Laurence A Angel et al.
The journal of physical chemistry. A, 110(35), 10392-10403 (2006-09-01)
Energy-resolved, competitive threshold collision-induced dissociation (TCID) methods are used to measure the gas-phase acidities of phenol, 3-methylphenol, 2,4,6-trimethylphenol, and ethanoic acid relative to hydrogen cyanide, hydrogen sulfide, and the hydroperoxyl radical using guided ion beam tandem mass spectrometry. The gas-phase
Claudio S Batista et al.
The Cochrane database of systematic reviews, (4)(4), CD006562-CD006562 (2010-04-16)
Genital warts are common and usually are harmless but can be painful and psychologically burdensome. Several local treatments can be used, including topical 5-Fluorouracil (5-FU). To determine the effectiveness and safety of 5-FU topical treatment for genital warts in nonimmunocompromised
G D Smith et al.
Acta crystallographica. Section D, Biological crystallography, 56(Pt 12), 1541-1548 (2000-11-28)
The structures of three R(6) human insulin hexamers have been determined. Crystals of monoclinic m-cresol-insulin, monoclinic resorcinol-insulin and rhombohedral m-cresol-insulin diffracted to 1. 9, 1.9 and 1.78 A, respectively, and have been refined to residuals of 0.195, 0.179 and 0.200

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service