Skip to Content
Merck
All Photos(1)

Key Documents

PRIH11AAF31F

Princeton BioMolecular Research, Inc.

5-chloro-2-(methylsulfonyl)-N-[2-(trifluoromethyl)phenyl]pyrimidine-4-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
A$1,129.41
1 G
A$1,731.00

Vendor SKU: SS-5272-500MG

A$1,129.41


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
500 MG
A$1,129.41
1 G
A$1,731.00

About This Item

Molecular Weight:
379.75
MDL number:

Vendor SKU: SS-5272-500MG

A$1,129.41


Ships in 21 days - from Aldrich Partner.

Iupac Name

5-chloro-2-(methylsulfonyl)-N-[2-(trifluoromethyl)phenyl]-4-pyrimidinecarboxamide

Inchi Code

1S/C13H9ClF3N3O3S/c1-24(22,23)12-18-6-8(14)10(20-12)11(21)19-9-5-3-2-4-7(9)13(15,16)17/h2-6H,1H3,(H,19,21)

InChI key

IKKYIYUGRDLJNV-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

Solid

Product Link

http://store.princetonbio.com/?product_id=PBMR278687

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service