Key Documents
MXQULTVP2
Millipore
Milliflex® Quantum system Validation Protocol for the combination with Milliflex® Oasis system
for use with Milliflex® Quantum Systems, for use with Milliflex Oasis® Systems
About This Item
UNSPSC Code:
41116500
NACRES:
ND.05
Pricing and availability is not currently available.
availability
available only in North America
compatibility
for use with Milliflex Oasis® Systems
for use with Milliflex® Quantum Systems
General description
Milliflex® Quantum Validation Protocol(Letter Format) is designed as a ready to use step by step guide to perform IQ and OQ activities of rapid microbial detection system using Milliflex® Quantum in combination with Milliflex®Oasis system. The protocol contains an exhaustive list of tests including test descriptions and test records. The protocol follows GMP, EP, USP and JP regulatory guidelines. Only available in North America.
Application
Milliflex® Quantum Validation Protocol is used to validate your rapid microbial detection system with ease and consistent performance while following regulatory guidelines.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Visit the Document Library
Articles
Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.