Skip to Content
Merck

Skip To

MXPHEAVHP

Milliflex® PLUS pump head VHP compatible

The Milliflex® PLUS pump head is a spare part of the Milliflex PLUS pump. It is suitable for use in isolators and is Vapor Hydrogen Peroxide (VHP) compatible.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 ea

Estimated to ship TODAYfromMerck Life Science BV/SRL

€727.00

About This Item

NACRES:
NB.79
UNSPSC Code:
23151818

€727.00


Estimated to ship TODAYDetails


Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

manufacturer/tradename

Milliflex®

technique(s)

Rapid Sterility Testing: suitable

input

sample type pharmaceutical(s)

application(s)

bioburden testing
pharmaceutical
sterility testing
water monitoring

compatibility

for use with MILLIFLEX®, standard for VHP

shipped in

ambient

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MMHEADMM1MXPFHUVHPMXPELEHUB
compatibility

for use with MILLIFLEX®, standard for VHP

compatibility

for use with Milliflex Oasis® filtration units, designed for MMSYSTMM1 pump

compatibility

for use with MILLIFLEX®, standard for VHP

compatibility

for use with MILLIFLEX®, standard for VHP

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

input

sample type pharmaceutical(s)

input

-

input

sample type pharmaceutical(s)

input

sample type pharmaceutical(s)

technique(s)

Rapid Sterility Testing: suitable

technique(s)

-

technique(s)

Rapid Sterility Testing: suitable

technique(s)

Rapid Sterility Testing: suitable

Quality Level

100

Quality Level

SP2

Quality Level

100

Quality Level

100

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex Oasis®

manufacturer/tradename

Milliflex®

manufacturer/tradename

PyroDetect, Milliflex®




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
MXPHEAVHP04053252552984

Questions

Reviews

No rating value

Active Filters