MPEVAR024
Microcon® Centrifugal Filters
Various MWCO, polyethersulfone membrane (Biomax)
Synonym(s):
Biomax polyethersulfone (PES) membrane centrifuge filter
Select a Size
€115.00
Estimated to ship on22 April 2025Details
Select a Size
About This Item
€115.00
Estimated to ship on22 April 2025Details
Recommended Products
material
polyacetal base (membrane support)
polycarbonate device top
polyethersulfone membrane (Biomax)
polypropylene tube (filtrate/concentrate)
silicone O-ring (medical grade)
description
Variety pack includes 4 of each filter size: 5K, 10K, 30K, 50K, 100K and 300K
sterility
non-sterile
product line
Microcon®
feature
holdup volume> 10 μL
pore size
10 kDa MWCO
100 kDa MWCO
30 kDa MWCO
300 kDa MWCO
5 kDa MWCO
50 kDa MWCO
General description
Simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1.5 mL tubes.
With the Biomax membrane, Microcon® filters offer:
- Reverse spin to maximize recovery, even in the smallest samples
- Convenient storage of filtrate or concentrated sample in standard microfuge tube
- Concentration factors up to 100X
Sample applications:
- Plant
Application
Features and Benefits
- High recovery for small volumes with reverse spin (concentration factor less than 20X)
- Biomax membrane
- Fast processing
Physical form
- Yellow
- Purple
- Pink
- Grey
- Green
- Amber
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service