Skip to Content
Merck
All Photos(1)

Documents

MABT1332

Sigma-Aldrich

Anti-Tspan-8 Antibody, clone 40/15-1B11-63-76-8

clone 40/15-1B11-63-76-8, from rat

Synonym(s):

Tetraspanin-8, Tspan-8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

40/15-1B11-63-76-8, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

monkey (based on 100% sequence homology), human (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Tspan8(216350)

General description

Tetraspanin-8 (UniProt: Q8R3G9; also known as Tspan8, Tspan-8) is encoded by the Tspan8 gene (Gene ID: 216350) in murine species. Tspan-8 is a multi-pass membrane protein of the tetraspanin family that is involved in the regulation of cell activation, growth, and motility. Members of this family have emerged as important regulators of expression levels, trafficking or post-translational modification of laminin-binding integrins. Tetraspanins are shown to associate with one another and cluster dynamically with a large variety of transmembrane and signal-transducing partners, forming specialized membrane microdomains called tetraspanin web. Tspan-8 is reported to be overexpressed during the progression of colorectal, liver, pancreatic and gastric cancers and its increased expression is shown to promote liver and lung metastasis. Overexpression of Tspan-8 has also been correlated with poor prognosis in human colorectal cancer. In metastatic pancreatic and colorectal carcinoma cell lines, PKC activation enhances the colocalization of Tspan-8 and Tspan CD151 with integrin alpha6beta 4 and subsequent internalization of this integrin-tetraspanin complex, resulting in decreased cell-matrix adhesion and increased cell migration. Tspan-8 knockdown in HT29 colon cancer cells is reported to significantly reduce cell migratory ability as a result of an upregulation of both integrin-dependent cell-matrix adhesion and calcium-dependent cell-cell adhesion. (Ref.: Kharbili, ME., et al. (2017). Oncotarget. 8(10); 17140-17155).

Specificity

Clone 40/15-1B11-63-76-8 is a rat monoclonal antibody that detects Tetraspanin-8. It targets an epitope with in the extracellular domain.

Immunogen

Epitope: extracellular domain
KLH-conjugated linear peptide corresponding to 10 amino acids from the extracellular domain of murine Tetraspanin-8.

Application

Anti-Tspan-8, clone 40/15-1B11-63-76-8 (Anti-Tspan8-A), Cat. No. MABT1332, is a rat monoclonal antibody that detects Tetraspanin-8 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected Tspan-8 in Immunohistochemistry applications (Fu, N.Y., et. al. (2017). Nat Cell Biol. 19(3):164-176).

Western Blotting Analysis: 4 µg/mL from a representative lot detected Tspan-8 in LIM1215 positive for Tspan8 and HEK293T cell lysates.
Research Category
Cell Structure

Quality

Evaluated by Immunohistochemistry (Paraffin) in mouse colon tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Tspan-8 in mouse colon tissue sections.

Target description

25.58 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibod yIgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service