MAB9251
Anti-Rubella Antibody, capsid, clone C8
ascites fluid, clone C8, Chemicon®
Synonym(s):
German Measles
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
ascites fluid
antibody product type
primary antibodies
clone
C8, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
immunofluorescence: suitable
isotype
IgM
shipped in
wet ice
Specificity
Specific for Rubella capsid protein. The antibody recognizes an epitope located within the amino terminus of the capsid protein located between amino acids 9 and 29.
Immunogen
Epitope: capsid
Application
Anti-Rubella Antibody, capsid, clone C8 detects level of Rubella & has been published & validated for use in IF.
IFA.
Western blot
Optimal working dilutions must be determined by end user.
Western blot
Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
Physical form
Ascites. Liquid.
Storage and Stability
Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid multiple freeze thaws.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Monoclonal antibody-defined epitope map of expressed rubella virus protein domains
Journal of virology, 65, 3986-3994 (1991)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service