HTS056M
ChemiSCREEN Membrane Preparation Recombinant Human FPRL1 (Formyl Peptide Receptor-Like 1)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
GPCR Class A
protein target FPRL1 / ALX / HM63
Quality Level
concentration
0.5 mg/mL
technique(s)
radioligand binding assay (RLBA): suitable
UniProt accession no.
Gene Information
human ... FPR2(2358)
General description
FUNCTION: SwissProt: P25090 # Low affinity receptor to N-formyl-methionyl peptides, which are powerful neutrophils chemotactic factors. Binding of FMLP to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. The activation of LXA4R could result in an anti-inflammatory outcome counteracting the actions of proinflammatory signals such as LTB4 (leukotriene B4).
SIZE: 351 amino acids; 38964 Da
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
TISSUE SPECIFICITY: Expressed abundantly in the lung and neutrophils. Also found in the spleen and testis.
SIMILARITY: SwissProt: P25090 ## Belongs to the G-protein coupled receptor 1 family.
SIZE: 351 amino acids; 38964 Da
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
TISSUE SPECIFICITY: Expressed abundantly in the lung and neutrophils. Also found in the spleen and testis.
SIMILARITY: SwissProt: P25090 ## Belongs to the G-protein coupled receptor 1 family.
Application
Radioligand binding assay and GTPγS binding.
Packaging
200 units
Quality
Kd for WKYMVm: 0.6 nM in a GTPγS binding assay
Physical form
Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membrane protein was adjusted to 0.5 mg/mL in 1 mL packaging buffer, rapidly frozen, and stored at -80°C.
Packaging method: Membrane protein was adjusted to 0.5 mg/mL in 1 mL packaging buffer, rapidly frozen, and stored at -80°C.
Storage and Stability
Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.
Analysis Note
ASSAY CONDITIONS: Membranes are permeabilized by addition of saponin to an equal concentration by mass, then mixed with [35S]-GTPγS (final concentration of 0.3 nM) in 20 mM HEPES, pH 7.4/100 mM NaCl/10 mM MgCl2/0.5 μM GDP in a nonbinding 96-well plate. Unlabeled WKYMVm is added to the final concentration indicated in Figure 1 (final volume 100 μL), and incubated for 30 min at 30°C. The binding reaction is transferred to an FB filter plate (Millipore MAHF B1H) pre-wetted with water, and washed 2 times (1 mL per well per wash) with cold 10 mM sodium phosphate, pH 7.4. The plate is dried and counted.
One vial contains enough membranes for at least 200 assays (units), where one unit is the amount of membrane that will yield greater than 1000 cpm specific WKYMVm stimulated [35S]-GTPγS binding when final concentration of 0.3nM [35S]-GTPγS was used.
One vial contains enough membranes for at least 200 assays (units), where one unit is the amount of membrane that will yield greater than 1000 cpm specific WKYMVm stimulated [35S]-GTPγS binding when final concentration of 0.3nM [35S]-GTPγS was used.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service