CC164
ECMatrix-111 E8 Laminin Substrate
Synonym(s):
ECMatrix-111 E8 Laminin Substrate, iMatrix-111
Select a Size
About This Item
Recommended Products
biological source
human
Quality Level
packaging
vial of 1050 μg
manufacturer/tradename
Millipore
Beta value
1) 6 X 175 µg ECMatrix-111 E8 Laminin Substrate (0.5 mg/mL in PBS). Expressed in CHO-S cells.
shipped in
2-8°C
storage temp.
2-8°C
General description
Application
- Purity (SDS-Page): <gt/> 95%
- Endotoxin Test: < 750 EU/mg
- Mycoplasma Test: Negative
- Sterility Test: Negative
- Integrin Binding Assay (kDa): < 10 nM
Features and Benefits
Storage and Stability
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service