Skip to Content
Merck
All Photos(2)

Documents

AB4332

Sigma-Aldrich

Anti-acetyl-MVH (Lys405) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Probable ATP-dependent RNA helicase DDX4, DEAD box protein 4, Mvh, Vasa homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

pig, human, mouse

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys405)

Gene Information

mouse ... Ddx4(13206)

General description

MVH (also known as DDX4 or Vasa) is a member of the DEAD family of proteins and is a probable ATP-dependent RNA helicase. MVH is mostly localized in testicular germ cells in the cytoplasm of spermatocytes. MVH is a high-copy extragenic suppressor gene. MVH is shown to be significantly decreased in spermatozoa of men that are infertile and may have an important role in male fertility and in stem cell development.

Specificity

This antibody specifically recognizes wild type MVH acetylated at Lys405, but not the Lys405 mutant.

Immunogen

Epitope: Acetylated Lys405
KLH-conjugated linear peptide corresponding to mouse MVH acetylated at Lys405.

Application

Anti-acetyl-MVH (Lys405) Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-MVH (Lys405) also known as Probable ATP-dependent RNA helicase DDX4, DEAD box protein & has been validated in DB, WB, ICC, PIA & IP.
Research Category
Stem Cell Research

Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blot/Immunoprecipitation Analysis: A representative lot detected Flag-tagged acetyl-MVH (Lys405) in the presence of Myc-HAT/p46.

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected acetyl-MVH (Lys405) in mouse germ cells (I. Nagamori, Department of Pharmacology, University of California, Irvine).

Quality

Evaluated by Dot Blot/Peptide Inhibition Assay in non-acetylated and acetylated MVH (Lys405).

Dot Blot/Peptide Inhibition Analysis: A 1:1,000 dilution of this antibody detected acetyl-MVH (Lys405) peptide.

Target description

76 kDa calculated

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Non-acetylated and acetylated MVH (Lys405)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service