Skip to Content
Merck
All Photos(1)

Documents

BSS-1006

Sigma-Aldrich

EmbryoMax® 1X Dulbecco′s Phosphate Buffered Saline w/o Ca++ & Mg++

1X, liquid, w/o Ca++ & Mg++ , EmbryoMax®

Synonym(s):

D-PBS w/o Ca ++/Mg ++, DPBS w/o Ca ++/Mg ++

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

product name

EmbryoMax® 1X Dulbecco′s Phosphate Buffered Saline w/o Ca++ & Mg++, The EmbryoMax 1X Dulbecco′s Phosphate Buffered Saline w/o Ca++ & Mg++ is available in a 1 L format and may be used for routine mouse embryonic stem cell culture applications.

form

liquid

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | stem cell: suitable

input

sample type neural stem cell(s)
sample type pancreatic stem cell(s)
sample type cardiac stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type hematopoietic stem cell(s)
sample type mesenchymal stem cell(s)
sample type induced pluripotent stem cell(s)
sample type epithelial cells
sample type: human embryonic stem cell(s)

General description

EmbryoMax 1X Dulbecco′s Phosphate Buffered Saline w/o Ca++ & Mg++

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

Mouse embryo media and embryo validated reagents for transgenic mouse embryo culture

Protocols

Stem Cell protocols for cryopreservation, thawing of cryopreserved stem cells and media preparation.

Related Content

Step-by-step protocol for generating apical-out human gut organoids for microbiome, ADME/Tox, viral and gastrointestinal related disease research. See the complete organoid culture protocol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service