5.08773
PPACK, 2HCl
InSolution, ≥88%
Synonym(s):
InSolution PPACK, 2HCl, D-Phenylalanyl-L-prolyl-L-arginine chloromethyl ketone, 2HCl, H-D-Phe-Pro-Arg-chloromethylketone, 2HCl, PPACK, dihydrochloride InSolution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥88% (HPLC)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
protect from light
storage temp.
−70°C
General description
D-Phenylalanyl-L-prolyl-L-arginine chloromethyl ketone (PPACK) is a potent and specific inhibitor of thrombin with ideal, unbiased properties for use as an anticoagulant in blood gas, electrolyte, and iCa analyses.
Biochem/physiol Actions
Reversible: no
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Physical form
A 25 mM (5 mg/357 µl) sterile-filtered solution of PPACK, 2HCl (Cat. No. 520222) in H₂O.
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.
Other Notes
Rosado, J. A., et al. 2002. J. Biol. Chem.277, 42157.
Odake S., et al. 1995. J. Enzyme Inhib.9, 17.
Arni, R.K., et al. 1993. Biochemistry32, 4727.
Yamashita, T., et al. 1993. Thromb. Res.69, 93.
Schmaier, A.H., et al. 1992. Thromb. Res.67, 479.
Collen, D., et al. 1982. J. Lab. Clin. Med.99, 76.
Kettner, C., and Shaw, E. 1981. Methods Enzymol.80, 826.
Kettner, C., and Shaw, E. 1979. Thromb. Res.14, 969.
Odake S., et al. 1995. J. Enzyme Inhib.9, 17.
Arni, R.K., et al. 1993. Biochemistry32, 4727.
Yamashita, T., et al. 1993. Thromb. Res.69, 93.
Schmaier, A.H., et al. 1992. Thromb. Res.67, 479.
Collen, D., et al. 1982. J. Lab. Clin. Med.99, 76.
Kettner, C., and Shaw, E. 1981. Methods Enzymol.80, 826.
Kettner, C., and Shaw, E. 1979. Thromb. Res.14, 969.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Clinical chemistry, 41(7), 1038-1041 (1995-07-01)
D-Phenylalanyl-L-prolyl-L-arginine chloromethyl ketone (PPACK), a selective thrombin inhibitor, was evaluated as an alternative anticoagulant to lithium heparin (LiHep) for blood gas and whole-blood electrolyte analyses. Initial studies in serum revealed a negative proportional bias for ionized calcium (iCa) concentration measured
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service