Skip to Content
Merck
All Photos(1)

Key Documents

20-108

Sigma-Aldrich

Assay Dilution Buffer I (ADBI)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€185.00

€185.00


Estimated to ship on25 February 2025Details



Select a Size

Change View
1 ML
€185.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.84

€185.00


Estimated to ship on25 February 2025Details


form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

Application

For use in kinase assays.
Research Category
All

Physical form

20mM MOPS, pH 7.2, 25mM β-glycerol phosphate, 5mM EGTA, 1mM sodium orthovanadate, 1mM dithiothreitol

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutagenicity testing of condensates of smoke from titanium dioxide/hexachloroethane and zinc/hexachloroethane pyrotechnic mixtures.
N Karlsson,I Fangmark,I Haggqvist,B Karlsson,L Rittfeldt,H Marchner
Mutation Research null
Limits for alkaline detoxification of dilute-acid lignocellulose hydrolysates.
Nils-Olof Nilvebrant,Per Persson,Anders Reimann,Filipe De Sousa,Lo Gorton,Leif J Jonsson
Applied Biochemistry and Biotechnology null
Permeation associated with three-phase-partitioning method on release of green fluorescent protein.
Thereza Christina Vessoni Penna,Eb Chiarini,Adalberto Pessoa Junior
Applied Biochemistry and Biotechnology null
Lactic acid production through cell-recycle repeated-batch bioreactor.
Hurok Oh,Young-Jung Wee,Jong-Sun Yun,Hwa-Won Ryu
Applied Biochemistry and Biotechnology null
Effect of lignocellulosic degradation compounds from steam explosion pretreatment on ethanol fermentation by thermotolerant yeast Kluyveromyces marxianus.
Jose Miguel Oliva,Felicia Saez,Ignacio Ballesteros,Alberto Gonzalez,Maria Jose Negro et al.
Applied Biochemistry and Biotechnology null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service