14-736-M
TAO2 Protein, active, 10 µg
Active, N-terminal 6His-tagged, recombinant, TAO2. TAO2 is a MAP3K that doubly phosphorylates & activates the MAP2Ks, MEK3 & MEK6. For use in Kinase Assays.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in Sf21 cells
Assay
84% (Coomassie blue staining, SDS-PAGE)
specific activity
1562 U/mg
mol wt
Mw 138.3 kDa
manufacturer/tradename
Chemicon®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
Gene Information
human ... TAOK2(9344)
General description
N-terminal 6His-tagged, recombinant, TAO2. TAO2 is a mitogen-activated protein kinase kinase kinase (MAP3K) that doubly phosphorylates and activates the MAP kinase kinases (MAP2Ks) MEK3 and MEK6.
Product Source: expressed in Sf21 cells
Application
Research Category
Metabolism
Metabolism
Research Sub Category
Cardiovascular Disease
Metabolic Disorders
Cardiovascular Disease
Metabolic Disorders
Biochem/physiol Actions
Protein Target: TAO2
Target Sub-Family: STE
Quality
Routinely evaluated by phosphorylation of 0.8 mg/ml myelin basic protein (MBP). Purity was assessed by SDS-PAGE and Coomassie blue staining using 3 μg of TAO2.
Unit Definition
Specific Activity: where one unit of TOA2 activity is defined as 1 nmol phosphate incorporated into 0.8 mg/mL myelin basic per minute at 30°C with a final ATP concentration of 100 μM.
Physical form
10 μg of enzyme in 100 μL of 50 mM Tris/HCl pH7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij 35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Ni2+/NTA-agarose
Storage and Stability
Stable for 6 months at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service