Skip to Content
Merck
All Photos(1)

Key Documents

14-521

Sigma-Aldrich

Arg Protein, active, human, 10 µg

Sign Into View Organizational & Contract Pricing

Select a Size

14-521
€412.00

€412.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
14-521
€412.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405

€412.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

description

protein target Arg

Quality Level

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... ABL2(27)

General description

FUNCTION: SwissProt: P42684 # Regulates cytoskeleton remodeling during cell differentiation, cell division and cell adhesion. Localizes to dynamic actin structures, and phosphorylates CRK and CRKL, DOK1, and other proteins controlling cytoskeleton dynamics.
COFACTOR: Magnesium or manganese.
SIZE: 1182 amino acids; 128343 Da
SUBCELLULAR LOCATION: Cytoplasm.
TISSUE SPECIFICITY: Widely expressed.
PTM: Phosphorylated upon DNA damage, probably by ATM or ATR.
SIMILARITY: SwissProt: P42684 ## Belongs to the protein kinase superfamily. Tyr protein kinase family. ABL subfamily. & Contains 1 protein kinase domain. & Contains 1 SH2 domain. & Contains 1 SH3 domain.

Quality

Routinely evaluated by phosphorylation of Abltide.

Physical properties

121.9 kDa

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service