Skip to Content
Merck
All Photos(1)

Key Documents

06-937

Sigma-Aldrich

Anti-phospho-STAT6 (Tyr641) Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€458.00

€458.00


Estimated to ship on14 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€458.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€458.00


Estimated to ship on14 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr641)

Gene Information

human ... STAT6(6778)

Specificity

phospho-STAT6

Immunogen

peptide corresponding to amino acids 631-645 of human STAT6 (C-PEQMGKDGRGpYVPAT)

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
This Anti-phospho-STAT6 (Tyr641) Antibody is validated for use in WB for the detection of phospho-STAT6 (Tyr641).

Quality

routinely evaluated by immunoblot on lysates of HepG2 cells, serum starved, then stimulated with IL-4

Target description

100kDa

Physical form

0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.
Format: Purified
Protein A purified

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 1 year after date of receipt.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F W Quelle et al.
Molecular and cellular biology, 15(6), 3336-3343 (1995-06-01)
By searching a database of expressed sequences, we identified a member of the signal transducers and activators of transcription (Stat) family of proteins. Human and murine full-length cDNA clones were obtained and sequenced. The sequence of the human cDNA was
Defective GATA-3 expression in Th2 LCR-deficient mice.
Soo Seok Hwang,Kiwan Kim,Gap Ryol Lee
Biochemical and biophysical research communications null
Giulia Veltri et al.
Cancers, 13(15) (2021-08-08)
Lymphoblastic lymphoma (LBL) is the second most common type of non-Hodgkin lymphoma in childhood, mainly of T cell origin (T-LBL). Although current treatment protocols allow a complete remission in 85% of cases, the second-line treatment overall survival for patients with
J Hou et al.
Science (New York, N.Y.), 265(5179), 1701-1706 (1994-09-16)
Interleukin-4 (IL-4) is an immunomodulatory cytokine secreted by activated T lymphocytes, basophils, and mast cells. It plays an important role in modulating the balance of T helper (Th) cell subsets, favoring expansion of the Th2 lineage relative to Th1. Imbalance
Tetsuya Kubota et al.
Nature communications, 9(1), 4863-4863 (2018-11-20)
M2a-subtype macrophage activation is known to be impaired in obesity, although the underlying mechanisms remain poorly understood. Herein, we demonstrate that, the IL-4/Irs2/Akt pathway is selectively impaired, along with decreased macrophage Irs2 expression, although IL-4/STAT6 pathway is maintained. Indeed, myeloid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service