Skip to Content
Merck
All Photos(1)

Key Documents

06-1130

Sigma-Aldrich

Anti-RNF168 Antibody

Synonym(s):

E3 ubiquitin-protein ligase RNF168, RING finger protein 168

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

rabbit

antibody form

affinity isolated antibody

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

General description

RNF168 is a ubiquitin ligand that binds to ubiquinated histone H2A and localizes to sites of DNA damage. RNF168-dependent chromatin modifications cause 53BP1 and BRCA1 to accumulated at the site. A mutation in RNF168 has been associated with RIDDLE syndrome, an immunodeficiency and radiosensitivity disorder. RNF168 is responsible for securing repair factors at sites of DNA damage. RNF168 has two MIU (motif interacting with ubiquitin) and one UMI (UIM- and MIU-related UBD) ubiquitin binding domains (UBDs) which are essential for proper localization of RNF168.

Specificity

This antibody recognizes RNF168 at the C-terminus.

Immunogen

Histidine-tagged recombinant protein corresponding to the C-terminus of human RNF168.

Application

Anti-RNF168 Antibody is a Rabbit Polyclonal Antibody for detection of RNF168 also known as E3 ubiquitin-protein ligase RNF168 & has been validated in WB.
Immunoprecipitation Analysis: A representative lot of this antibody was used to immunoprecipitate RNF168 from whole cell lysates (Zhao, H., et al. (2014) Cell Cycle. 13(11):1–11).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: 0.1 μg/mL of this antibody detected RNF168 on 10 μg of Jurkat cell lysate.

Target description

~79 kDa observed.

Physical form

Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate

Other Notes

Concentration:Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongchang Zhao et al.
Cell cycle (Georgetown, Tex.), 13(11), 1777-1787 (2014-04-16)
Timely and proper cellular response to DNA damage is essential for maintenance of genome stability and integrity. B-cell lymphoma/leukemia 10 (BCL10) facilitates ubiquitination of NEMO in the cytosol, activating NFκB signaling. Translocation and/or point mutations of BCL10 associate with mucosa-associated
Katherine N Choe et al.
EMBO reports, 17(6), 874-886 (2016-05-06)
Defects in DNA replication, DNA damage response, and DNA repair compromise genomic stability and promote cancer development. In particular, unrepaired DNA lesions can arrest the progression of the DNA replication machinery during S-phase, causing replication stress, mutations, and DNA breaks.
Lianzhong Zhang et al.
Genomics, proteomics & bioinformatics, 16(6), 428-438 (2018-12-12)
DNA damage response (DDR) is essential for maintaining genome stability and protecting cells from tumorigenesis. Ubiquitin and ubiquitin-like modifications play an important role in DDR, from signaling DNA damage to mediating DNA repair. In this report, we found that the
Kathy Shire et al.
Journal of cell science, 129(3), 580-591 (2015-12-18)
Promyelocytic leukemia (PML) protein forms the basis of PML nuclear bodies (PML NBs), which control many important processes. We have screened an shRNA library targeting ubiquitin pathway proteins for effects on PML NBs, and identified RNF8 and RNF168 DNA-damage response
Tina Gruosso et al.
EMBO molecular medicine, 8(5), 527-549 (2016-03-24)
Anti-cancer drugs often increase reactive oxygen species (ROS) and cause DNA damage. Here, we highlight a new cross talk between chronic oxidative stress and the histone variant H2AX, a key player in DNA repair. We observe that persistent accumulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service