Skip to Content
Merck
All Photos(1)

Key Documents

MAT239354893

Matrix Scientific

Ethyl 1-{[(tert-butoxycarbonyl)amino]sulfonyl}-4-piperidinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
336.41
MDL number:

mp

79-81

Iupac Name

ethyl 1-{[(tert-butoxycarbonyl)amino]sulfonyl}-4-piperidinecarboxylate

Inchi Code

1S/C13H24N2O6S/c1-5-20-11(16)10-6-8-15(9-7-10)22(18,19)14-12(17)21-13(2,3)4/h10H,5-9H2,1-4H3,(H,14,17)

InChI key

HWLHFRDIBQJGNI-UHFFFAOYSA-N

Purity

95%

Country of Origin

US

Product Link

https://www.matrixscientific.com/047811.html

Melting Point

79-81

Pictograms

MSDS

https://www.matrixscientific.com/047811.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service