KEY465193426
Key Organics / BIONET
methyl (2E)-3-(quinolin-3-yl)prop-2-enoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl (2E)-3-(3-quinolinyl)-2-propenoate
Inchi Code
1S/C13H11NO2/c1-16-13(15)7-6-10-8-11-4-2-3-5-12(11)14-9-10/h2-9H,1H3/b7-6+
InChI key
GVXTZYRMZZIIKT-VOTSOKGWSA-N
Purity
90%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/1R-0147
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/1R-0147
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service