KEY240887140
Key Organics / BIONET
1-bromo-4-chloro-2-nitro-5-(trifluoromethyl)benzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
58 - 60
Iupac Name
1-bromo-4-chloro-2-nitro-5-(trifluoromethyl)benzene
Inchi Code
1S/C7H2BrClF3NO2/c8-4-1-3(7(10,11)12)5(9)2-6(4)13(14)15/h1-2H
InChI key
COXWQBVLBUBRPG-UHFFFAOYSA-N
Purity
95%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/KD-0771
Melting Point
58 - 60
Signal Word
Warning
Hazard Codes
Precautionary statements
MSDS
https://www.keyorganics.net/sds/KD-0771
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service