Skip to Content
Merck
All Photos(1)

Key Documents

KEY187620340

Key Organics / BIONET

5-chloro-6-cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
208.65
MDL number:

Iupac Name

5-chloro-6-cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine

Inchi Code

1S/C9H9ClN4/c10-6-3-5-8(11)13-14-9(5)12-7(6)4-1-2-4/h3-4H,1-2H2,(H3,11,12,13,14)

InChI key

GQCSDBLOEWYZNU-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/AD-5013

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/AD-5013


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service