Skip to Content
Merck
All Photos(1)

Key Documents

KEY040898029

Key Organics / BIONET

1-(5-nitropyridin-2-yl)piperidine-4-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€175.30
1 G
€248.80
2 G
€446.80
5 G
€941.80
10 G
€1,634.80
20 G
€3,020.80

Vendor SKU: 2Y-0705-500MG

€175.30


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
500 MG
€175.30
1 G
€248.80
2 G
€446.80
5 G
€941.80
10 G
€1,634.80
20 G
€3,020.80

About This Item

CAS Number:
Molecular Weight:
251.24
MDL number:

Vendor SKU: 2Y-0705-500MG

€175.30


Ships in 21 days - from Aldrich Partner.

mp

186 - 188

Iupac Name

1-(5-nitro-2-pyridinyl)-4-piperidinecarboxylic acid

Inchi Code

1S/C11H13N3O4/c15-11(16)8-3-5-13(6-4-8)10-2-1-9(7-12-10)14(17)18/h1-2,7-8H,3-6H2,(H,15,16)

InChI key

RKRUADFRDLUFHR-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/2Y-0705

Melting Point

186 - 188

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/2Y-0705


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service