JWPH3249D0B2
J & W Pharmlab LLC
(5-Methoxy-1H-indazol-3-yl)-acetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(5-methoxy-1H-indazol-3-yl)acetic acid
Inchi Code
1S/C10H10N2O3/c1-15-6-2-3-8-7(4-6)9(12-11-8)5-10(13)14/h2-4H,5H2,1H3,(H,11,12)(H,13,14)
InChI key
ALDYNZAFBDWYOF-UHFFFAOYSA-N
Purity
97%
Country of Origin
US
Physical Form
White Solid
Product Link
https://www.jwpharmlab.com/home/product/45R0044
MSDS
https://www.jwpharmlab.com/home/product/45R0044
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service